missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylurea, 97%

Catalog No. AAL0207906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0207906 5 g
AAL0207914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0207906 Supplier Thermo Scientific Chemicals Supplier No. L0207906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 623-76-7 | C5H12N2O | 116.16 g/mol

N,N'-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylurea is used for synthesis of caffeine, theophylline, pharma chemicals, textile aids

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 623-76-7
Molecular Formula C5H12N2O
Molecular Weight (g/mol) 116.16
MDL Number MFCD00009028
InChI Key ZWAVGZYKJNOTPX-UHFFFAOYSA-N
Synonym n,n'-diethylurea, sym-diethylurea, urea, n,n'-diethyl, urea, 1,3-diethyl, urea, n,n'-diethyl 9ci, sym-n,n'-diethylurea, n-ethyl ethylamino carboxamide, urea,3-diethyl, urea,n'-diethyl, 1,3-diethyl-urea
PubChem CID 12194
IUPAC Name 1,3-diethylurea
SMILES CCNC(=O)NCC

Specifications

Melting Point 109°C to 113°C
Linear Formula CH3CH2NHCONHCH2CH3
Quantity 5 g
Beilstein 1744741
Solubility Information Soluble in water.
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material N,N'-Diethylurea
EINECSNumber 210-811-3
RTECSNumber YS9354000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only