accessibility menu, dialog, popup

UserName

H3CHNOHNCH3

CAS RN 623-76-7

IUPAC Name: 1,3-diethylurea
Synonyms: urea, N,N'-diethyl-
Molecular Weight (g/mol): 116.16
Molecular Formula: C5H12N2O
InChi Key: ZWAVGZYKJNOTPX-UHFFFAOYSA-N
SMILES: CCNC(=O)NCC
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)