missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylsalicylamide, 97%, Thermo Scientific™

Chemical Identifiers

CAS 19311-91-2
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.24
InChI Key ZVYXEXAXXWINEH-UHFFFAOYSA-N
Synonym n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide
PubChem CID 88006
IUPAC Name N,N-diethyl-2-hydroxybenzamide
SMILES CCN(CC)C(=O)C1=CC=CC=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC437250050
SDS View Documents
Thermo Scientific Chemicals
437250050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC437250250
SDS View Documents
Thermo Scientific Chemicals
437250250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

19311-91-2
193.24
n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide
N,N-diethyl-2-hydroxybenzamide
C11H15NO2
ZVYXEXAXXWINEH-UHFFFAOYSA-N
88006
CCN(CC)C(=O)C1=CC=CC=C1O

Specifications

19311-91-2
100.0
96% min. (GC)
C11H15NO2
n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide
CCN(CC)C(=O)C1=CC=CC=C1O
193.24
193.24
N, N-Diethylsalicylamide
96.0
Authentic
Glass bottle
5g
ZVYXEXAXXWINEH-UHFFFAOYSA-N
N,N-diethyl-2-hydroxybenzamide
88006
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-955-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only