missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylhydroxylamine (ca. 85% in Water, ca. 8.6mol/L), TCI America™
Supplier: TCI America D1178500ML
Specifications
| N,N-Diethylhydroxylamine (ca. 85% in Water, ca. 8.6mol/L) | |
| Yellow | |
| MFCD00002126 | |
| 1993 | |
| FVCOIAYSJZGECG-UHFFFAOYSA-N | |
| N,N-diethylhydroxylamine | |
| 19463 | |
| Liquid |
| 3710-84-7 | |
| C4H11NO | |
| 500 mL | |
| diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine | |
| CCN(CC)O | |
| 89.138 | |
| 89.14 |
Chemical Identifiers
| 3710-84-7 | |
| 89.138 | |
| FVCOIAYSJZGECG-UHFFFAOYSA-N | |
| 19463 | |
| CCN(CC)O |
| C4H11NO | |
| MFCD00002126 | |
| diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine | |
| N,N-diethylhydroxylamine |
Safety and Handling
EINECSNumber : (2)-0190
RTECSNumber : NC3500000
TSCA : Yes