missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylhydroxylamine (ca. 85% in Water, ca. 8.6mol/L), TCI America™

Catalog No. p-7126628
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D117825ML 25 mL
D1178500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D117825ML Supplier TCI America Supplier No. D117825ML
Only null left

Chemical Identifiers

CAS 3710-84-7
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00002126
InChI Key FVCOIAYSJZGECG-UHFFFAOYSA-N
Synonym diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
PubChem CID 19463
IUPAC Name N,N-diethylhydroxylamine
SMILES CCN(CC)O

Specifications

Color Yellow
Quantity 25 mL
UN Number 1993
Formula Weight 89.14
Physical Form Liquid
Chemical Name or Material N,N-Diethylhydroxylamine (ca. 85% in Water, ca. 8.6mol/L)
EINECSNumber (2)-0190
RTECSNumber NC3500000
TSCA Yes