missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylglycine Methyl Ester 98.0+%, TCI America™

Catalog No. p-7126730
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D144325G 25 g
D1443500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D144325G Supplier TCI America Supplier No. D144325G

Chemical Identifiers

CAS 30280-35-4
Molecular Formula C7H15NO2
Molecular Weight (g/mol) 145.20
MDL Number MFCD00059368
InChI Key YLZLHVWLTRZOJH-UHFFFAOYSA-N
PubChem CID 559451
IUPAC Name methyl 2-(diethylamino)acetate
SMILES CCN(CC)CC(=O)OC

Specifications

Color Yellow
Boiling Point 86°C
Quantity 25 g
UN Number 1993
Formula Weight 145.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylglycine Methyl Ester
TSCA No
Recommended Storage Refrigerator