accessibility menu, dialog, popup

UserName

H3CNCH3OOCH3

CAS RN 30280-35-4

IUPAC Name: methyl 2-(diethylamino)acetate
Synonyms: methyl 2-(diethylamino)acetate
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: YLZLHVWLTRZOJH-UHFFFAOYSA-N
SMILES: CCN(CC)CC(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results