missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Diethylethylenediamine 95.0+%, TCI America™
$89.48 - $242.33
Chemical Identifiers
| CAS | 111-74-0 |
|---|---|
| Molecular Formula | C6H16N2 |
| Molecular Weight (g/mol) | 116.21 |
| MDL Number | MFCD00009033 |
| InChI Key | CJKRXEBLWJVYJD-UHFFFAOYSA-N |
| Synonym | n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide |
| PubChem CID | 67105 |
| ChEBI | CHEBI:182290 |
| IUPAC Name | ethyl[2-(ethylamino)ethyl]amine |
| SMILES | CCNCCNCC |
Chemical Identifiers
| 111-74-0 | |
| 116.21 | |
| CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
| 67105 | |
| ethyl[2-(ethylamino)ethyl]amine |
| C6H16N2 | |
| MFCD00009033 | |
| n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
| CHEBI:182290 | |
| CCNCCNCC |
Specifications
| 111-74-0 | |
| 150°C | |
| MFCD00009033 | |
| 2685 | |
| CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
| ethyl[2-(ethylamino)ethyl]amine | |
| 67105 | |
| 116.21 | |
| Liquid |
| Yellow | |
| C6H16N2 | |
| 5 mL | |
| n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
| CCNCCNCC | |
| 116.21 | |
| CHEBI:182290 | |
| ≥95.0% (GC) | |
| N,N′-Diethylethylenediamine |
Safety and Handling
TSCA : Yes