Learn More
N,N'-Diethylethylenediamine, 96%
CAS: 111-74-0 | C6H16N2 | 116.21 g/mol
Supplier: Thermo Scientific Chemicals A1711322
Description
N,N'-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N'-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N′-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N′-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.
Solubility
Miscible with water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide and strong oxidizing agents.
Specifications
| N,N'-Diethylethylenediamine | |
| 0.811 | |
| 33°C (92°F) | |
| 1.4326 | |
| 100 g | |
| Air Sensitive | |
| Miscible with water. | |
| CCNCCNCC | |
| 116.21 | |
| CHEBI:182290 | |
| 96% |
| 111-74-0 | |
| 152°C to 154°C | |
| C6H16N2 | |
| MFCD00009033 | |
| UN2685 | |
| n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
| CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
| N,N'-diethylethane-1,2-diamine | |
| 67105 | |
| 116.21 |
Chemical Identifiers
| 111-74-0 | |
| 116.21 | |
| CJKRXEBLWJVYJD-UHFFFAOYSA-N | |
| 67105 | |
| N,N'-diethylethane-1,2-diamine |
| C6H16N2 | |
| MFCD00009033 | |
| n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide | |
| CHEBI:182290 | |
| CCNCCNCC |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: N,N-DIETHYLETHYLENEDIAMINE
EINECSNumber : 203-902-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only