missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylbenzylamine 98.0+%, TCI America™
$83.89 - $83.89
Chemical Identifiers
| CAS | 772-54-3 |
|---|---|
| Molecular Formula | C11H17N |
| Molecular Weight (g/mol) | 163.26 |
| MDL Number | MFCD00026731 |
| InChI Key | ZWRDBWDXRLPESY-UHFFFAOYSA-N |
| Synonym | N-Benzyldiethylamine |
| PubChem CID | 61211 |
| IUPAC Name | benzyldiethylamine |
| SMILES | CCN(CC)CC1=CC=CC=C1 |
Chemical Identifiers
| 772-54-3 | |
| 163.26 | |
| ZWRDBWDXRLPESY-UHFFFAOYSA-N | |
| 61211 | |
| CCN(CC)CC1=CC=CC=C1 |
| C11H17N | |
| MFCD00026731 | |
| N-Benzyldiethylamine | |
| benzyldiethylamine |
Specifications
| 772-54-3 | |
| 216°C | |
| MFCD00026731 | |
| 2735 | |
| ZWRDBWDXRLPESY-UHFFFAOYSA-N | |
| benzyldiethylamine | |
| 61211 | |
| ≥98.0% (GC,T) | |
| N,N-Diethylbenzylamine |
| Yellow | |
| C11H17N | |
| 25 mL | |
| N-Benzyldiethylamine | |
| CCN(CC)CC1=CC=CC=C1 | |
| 163.26 | |
| 163.26 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0350
TSCA : No
Recommended Storage : Refrigerator