missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylaniline Borane 98.0+%, TCI America™
Supplier: TCI America D2581100G
Specifications
| N,N-Diethylaniline Borane | |
| -40°C | |
| C10H15BN | |
| 100 g | |
| n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex | |
| [B].CCN(CC)C1=CC=CC=C1 | |
| 160.05 | |
| 163.07 | |
| Liquid |
| 13289-97-9 | |
| Yellow | |
| MFCD00013187 | |
| 3148 | |
| KHYAFFAGZNCWPT-UHFFFAOYSA-N | |
| N,N-diethylaniline boron | |
| 6335292 | |
| ≥98.0% (T) |
Chemical Identifiers
| 13289-97-9 | |
| 160.05 | |
| KHYAFFAGZNCWPT-UHFFFAOYSA-N | |
| 6335292 | |
| [B].CCN(CC)C1=CC=CC=C1 |
| C10H15BN | |
| MFCD00013187 | |
| n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex | |
| N,N-diethylaniline boron |
Safety and Handling
TSCA : Yes