missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylaniline Borane 98.0+%, TCI America™

Catalog No. p-7127381
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D258125G 25 g
D2581100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D258125G Supplier TCI America Supplier No. D258125G
Only null left

Chemical Identifiers

CAS 13289-97-9
Molecular Formula C10H15BN
Molecular Weight (g/mol) 160.05
MDL Number MFCD00013187
InChI Key KHYAFFAGZNCWPT-UHFFFAOYSA-N
Synonym n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
PubChem CID 6335292
IUPAC Name N,N-diethylaniline boron
SMILES [B].CCN(CC)C1=CC=CC=C1

Specifications

Melting Point -40°C
Color Yellow
Quantity 25 g
UN Number 3148
Formula Weight 163.07
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material N,N-Diethylaniline Borane