missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-N'-oleoylethylenediamine 97.0+%, TCI America™
$63.38 - $315.03
Chemical Identifiers
| CAS | 13282-67-2 |
|---|---|
| Molecular Formula | C24H48N2O |
| Molecular Weight (g/mol) | 380.66 |
| MDL Number | MFCD00027344 |
| InChI Key | WCSBDRIEINNXNW-BUHFOSPRSA-N |
| PubChem CID | 6146519 |
| IUPAC Name | (9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide |
| SMILES | CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC |
Chemical Identifiers
| 13282-67-2 | |
| 380.66 | |
| WCSBDRIEINNXNW-BUHFOSPRSA-N | |
| (9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide |
| C24H48N2O | |
| MFCD00027344 | |
| 6146519 | |
| CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC |
Specifications
| 13282-67-2 | |
| C24H48N2O | |
| 5 g | |
| WCSBDRIEINNXNW-BUHFOSPRSA-N | |
| (9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide | |
| 6146519 | |
| ≥97.0% (T) | |
| N,N-Diethyl-N′-oleoylethylenediamine |
| Brown-Yellow | |
| MFCD00027344 | |
| N′C-Oleoyl-N,N-diethylethylenediamine | |
| CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC | |
| 380.66 | |
| 380.66 | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-0276
TSCA : No