CAS RN 13282-67-2
CAS RN 13282-67-2
IUPAC Name:
(9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide
Synonyms:
(9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide
Molecular Weight (g/mol):
380.66
Molecular Formula:
C24H48N2O
InChi Key:
WCSBDRIEINNXNW-BUHFOSPRSA-N
SMILES:
CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC
1
–
1
of
1
results
1
N,N-Diethyl-N'-oleoylethylenediamine 97.0+%, TCI Americaâ„¢
CAS: 13282-67-2 Molecular Formula: C24H48N2O Molecular Weight (g/mol): 380.66 MDL Number: MFCD00027344 InChI Key: WCSBDRIEINNXNW-BUHFOSPRSA-N PubChem CID: 6146519 IUPAC Name: (9E)-N-[2-(diethylamino)ethyl]octadec-9-enamide SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)NCCN(CC)CC