missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-m-toluamide 98.0+%, TCI America™

Catalog No. D009725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D009725ML 25 mL
D0097500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D009725ML Supplier TCI America Supplier No. D009725ML

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00009046
InChI Key MMOXZBCLCQITDF-UHFFFAOYSA-N
Synonym deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
PubChem CID 4284
ChEBI CHEBI:7071
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)C1=CC(=CC=C1)C

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 191.27
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-m-toluamide
EINECSNumber (3)-1321&(3)-1677
RTECSNumber XS3675000
TSCA Yes