missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-3-methylbenzamide, 97%

Catalog No. AAA1748636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1748636 500 g
AAA1748622 100 g
AAA174860E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1748636 Supplier Thermo Scientific Chemicals Supplier No. A1748636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-62-3 | C12H17NO | 191.274 g/mol

It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

Solubility
Soluble in water.

Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00009046
InChI Key MMOXZBCLCQITDF-UHFFFAOYSA-N
Synonym deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
PubChem CID 4284
ChEBI CHEBI:7071
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)C1=CC(=CC=C1)C

Specifications

Density 0.997
Boiling Point 110°C to 112°C (1 mmHg)
Flash Point 145°C (293°F)
Refractive Index 1.523
Quantity 500 g
Beilstein 2046711
Merck Index 14,2856
Solubility Information Soluble in water.
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-3-methylbenzamide
Show More Show Less
Hazard Category H302-H315-H319-H336-H373
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
EINECSNumber 205-149-7
RTECSNumber XS3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only