missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethyl-1,4-phenylenediamine 98.0+%, TCI America™
$217.54 - $3143.80
Chemical Identifiers
| CAS | 93-05-0 |
|---|---|
| Molecular Formula | C10H16N2 |
| Molecular Weight (g/mol) | 164.252 |
| MDL Number | MFCD00007861 |
| InChI Key | QNGVNLMMEQUVQK-UHFFFAOYSA-N |
| Synonym | n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline |
| PubChem CID | 7120 |
| IUPAC Name | 4-N,4-N-diethylbenzene-1,4-diamine |
| SMILES | CCN(CC)C1=CC=C(C=C1)N |
Chemical Identifiers
| 93-05-0 | |
| 164.252 | |
| QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
| 7120 | |
| CCN(CC)C1=CC=C(C=C1)N |
| C10H16N2 | |
| MFCD00007861 | |
| n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline | |
| 4-N,4-N-diethylbenzene-1,4-diamine |
Specifications
| 93-05-0 | |
| 262°C | |
| MFCD00007861 | |
| 2922 | |
| QNGVNLMMEQUVQK-UHFFFAOYSA-N | |
| 4-N,4-N-diethylbenzene-1,4-diamine | |
| 7120 | |
| ≥98.0% (GC,T) | |
| N,N-Diethyl-1,4-phenylenediamine |
| Brown-Yellow | |
| C10H16N2 | |
| 25 g | |
| n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline | |
| CCN(CC)C1=CC=C(C=C1)N | |
| 164.252 | |
| 164.25 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-0243
RTECSNumber : SS9275000
TSCA : Yes
Recommended Storage : Freezer