missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America D0518500G

 View more versions of this product

Catalog No. D0518500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N,N-Diethyl-1,4-phenylenediamine
Brown-Yellow
C10H16N2
500 g
n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
CCN(CC)C1=CC=C(C=C1)N
164.252
164.25
Liquid
93-05-0
262°C
MFCD00007861
2922
QNGVNLMMEQUVQK-UHFFFAOYSA-N
4-N,4-N-diethylbenzene-1,4-diamine
7120
≥98.0% (GC,T)

Chemical Identifiers

93-05-0
164.252
QNGVNLMMEQUVQK-UHFFFAOYSA-N
7120
CCN(CC)C1=CC=C(C=C1)N
C10H16N2
MFCD00007861
n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
4-N,4-N-diethylbenzene-1,4-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0243

RTECSNumber : SS9275000

TSCA : Yes

Recommended Storage : Freezer

SDS
Documents

Documents

Promotions

Promotions