missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,4-phenylenediamine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7126264
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D05195G 5 g
D051925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D05195G Supplier TCI America Supplier No. D05195G

Chemical Identifiers

CAS 16713-15-8
Molecular Formula C10H18Cl2N2
Molecular Weight (g/mol) 237.168
MDL Number MFCD00060227
InChI Key NHPCFKHZILUMDE-UHFFFAOYSA-N
Synonym 4-Amino-N,N-diethylaniline Dihydrochloride
PubChem CID 19799983
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;dihydrochloride
SMILES CCN(CC)C1=CC=C(C=C1)N.Cl.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 237.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Diethyl-1,4-phenylenediamine Dihydrochloride
EINECSNumber (3)-0243
TSCA Yes