accessibility menu, dialog, popup

UserName

HClCH3NCH3NH2

CAS RN 16713-15-8

IUPAC Name: hydrogen N1,N1-diethylbenzene-1,4-diamine chloride
Synonyms: hydrogen N,N-diethyl-1,4-benzenediamine chloride
Molecular Weight (g/mol): 200.71
Molecular Formula: C10H17ClN2
InChi Key: XTBFKMDOQMQYPP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCN(CC)C1=CC=C(N)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results