missing translation for 'onlineSavingsMsg'
Learn More

N,N-Didodecylmethylamine 85.0+%, TCI America™

Catalog No. p-7127674
Change view
Click to view available options
Quantity:
25 g
400 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D333825G 25 g
D3338400G 400 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D333825G Supplier TCI America Supplier No. D333825G
Only null left

Chemical Identifiers

CAS 2915-90-4
Molecular Formula C25H53N
Molecular Weight (g/mol) 367.706
InChI Key UWHRNIXHZAWBMF-UHFFFAOYSA-N
Synonym N-Methyldidodecylamine
PubChem CID 76205
IUPAC Name N-dodecyl-N-methyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)CCCCCCCCCCCC

Specifications

Melting Point 16°C
Color Yellow
Boiling Point 204°C
Quantity 25 g
Formula Weight 367.71
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Didodecylmethylamine
EINECSNumber (2)-0176
TSCA Yes