accessibility menu, dialog, popup

UserName

CAS RN 2915-90-4

H3CNCH3CH3

CAS RN 2915-90-4

IUPAC Name: didodecyl(methyl)amine
Synonyms: didodecyl(methyl)amine
Molecular Weight (g/mol): 367.71
Molecular Formula: C25H53N
InChi Key: UWHRNIXHZAWBMF-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN(C)CCCCCCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results