missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexyl-4-morpholinecarboxamidine 98.0+%, TCI America™

Catalog No. D37925G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37925G 5 g
D379225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37925G Supplier TCI America Supplier No. D37925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4975-73-9
Molecular Formula C17H31N3O
Molecular Weight (g/mol) 293.455
MDL Number MFCD00063252
InChI Key OZNYZQOTXQSUJM-UHFFFAOYSA-N
Synonym n,n'-dicyclohexyl-4-morpholinecarboxamidine, n,n'-dicyclohexyl-4-morpholine carboxamidine, n,n'-dicyclohexyl-1-morpholinoformamidine, n,n'-dicyclohexylmorpholine-4-carboxamidine, 4-morpholinecarboximidamide, n,n'-dicyclohexyl, formamidine, n,n'-dicyclohexyl-1-morpholino, n,n'-dicyclohexyl-4-morpholine-carboxamidine, z-n,n'-dicyclohexylmorpholine-4-carboximidamide, 4-n,n'-dicyclohexylamidino morpholine, n,n'-dicyclohexyl-4-morpholinecarboximidamide
PubChem CID 21079
IUPAC Name N,N'-dicyclohexylmorpholine-4-carboximidamide
SMILES C1CCC(CC1)NC(=NC2CCCCC2)N3CCOCC3

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 5 g
Formula Weight 293.46
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dicyclohexyl-4-morpholinecarboxamidine
RTECSNumber LQ4566500
TSCA No