accessibility menu, dialog, popup

UserName

CAS RN 4975-73-9

IUPAC Name: N,N'-dicyclohexylmorpholine-4-carboximidamide
Synonyms: N,N'-dicyclohexylmorpholine-4-carboximidamide
Molecular Weight (g/mol): 293.46
Molecular Formula: C17H31N3O
InChi Key: OZNYZQOTXQSUJM-UHFFFAOYSA-N
SMILES: C1CCC(CC1)NC(=NC1CCCCC1)N1CCOCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results