missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dicyclohexyl-1,2-ethanediamine Hydrate 98.0+%, TCI America™
Supplier: TCI America D451825G
Specifications
| N,N′-Dicyclohexyl-1,2-ethanediamine Hydrate | |
| 95°C | |
| 312°C | |
| 25 g | |
| 1,2-Bis(cyclohexylamino)ethane | |
| C1CCC(CC1)NCCNC2CCCCC2 | |
| 224.392 | |
| 224.39 | |
| Crystalline Powder |
| 4013-98-3 | |
| White | |
| C14H28N2 | |
| 3259 | |
| JMRWVHXGVONXEZ-UHFFFAOYSA-N | |
| N,N'-dicyclohexylethane-1,2-diamine | |
| 273886 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 4013-98-3 | |
| 224.392 | |
| 1,2-Bis(cyclohexylamino)ethane | |
| N,N'-dicyclohexylethane-1,2-diamine |
| C14H28N2 | |
| JMRWVHXGVONXEZ-UHFFFAOYSA-N | |
| 273886 | |
| C1CCC(CC1)NCCNC2CCCCC2 |
Safety and Handling
TSCA : No