accessibility menu, dialog, popup

UserName

NHHN

CAS RN 4013-98-3

IUPAC Name: N1,N2-dicyclohexylethane-1,2-diamine
Synonyms: N1,N2-dicyclohexylethane-1,2-diamine
Molecular Weight (g/mol): 224.39
Molecular Formula: C14H28N2
InChi Key: JMRWVHXGVONXEZ-UHFFFAOYSA-N
SMILES: C(CNC1CCCCC1)NC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results