missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexyl-1,2-ethanediamine Hydrate 98.0+%, TCI America™

Catalog No. D45185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45185G 5 g
D451825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45185G Supplier TCI America Supplier No. D45185G

Chemical Identifiers

CAS 4013-98-3
Molecular Formula C14H28N2
Molecular Weight (g/mol) 224.392
InChI Key JMRWVHXGVONXEZ-UHFFFAOYSA-N
Synonym 1,2-Bis(cyclohexylamino)ethane
PubChem CID 273886
IUPAC Name N,N'-dicyclohexylethane-1,2-diamine
SMILES C1CCC(CC1)NCCNC2CCCCC2

Specifications

Melting Point 95°C
Color White
Boiling Point 312°C
Quantity 5 g
UN Number 3259
Formula Weight 224.39
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dicyclohexyl-1,2-ethanediamine Hydrate