missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicarbobenzoxy-L-cystine 98.0+%, TCI America™

Catalog No. D11971G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11971G 1 g
D11975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D11971G Supplier TCI America Supplier No. D11971G
Only null left

Chemical Identifiers

CAS 6968-11-2
Molecular Formula C22H24N2O8S2
Molecular Weight (g/mol) 508.56
MDL Number MFCD00022030
InChI Key PTRQEEVKHMDMCF-UHFFFAOYNA-N
Synonym z-cys-oh 2, cbz-cys-oh 2, di-z-l-cystine, n,n'-dicarbobenzoxy-l-cystine, n,n'-di-cbz-l-cystine, 2r-2-benzyloxy carbonyl amino-3-2r-2-benzyloxy carbonyl amino-2-carboxyethyl disulfanyl propanoic acid, n,n'-dibenzyloxycarbonyl-l-cystine, n,n'-bis benzyloxycarbonyl-l-cystine, ambotzzaa1190, pubchem19022
PubChem CID 385972
IUPAC Name (2R)-3-[[(2R)-2-carboxy-2-(phenylmethoxycarbonylamino)ethyl]disulfanyl]-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CSSCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 119°C
Color White
Quantity 1 g
Formula Weight 508.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Dicarbobenzoxy-L-cystine