accessibility menu, dialog, popup

UserName

OHOSSHNOOHOONHOO

CAS RN 6968-11-2

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-3-[(2-{[(benzyloxy)carbonyl]amino}-2-carboxyethyl)disulfanyl]propanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-3-[(2-{[(benzyloxy)carbonyl]amino}-2-carboxyethyl)disulfanyl]propanoic acid
Molecular Weight (g/mol): 508.56
Molecular Formula: C22H24N2O8S2
InChi Key: PTRQEEVKHMDMCF-UHFFFAOYNA-N
SMILES: OC(=O)C(CSSCC(NC(=O)OCC1=CC=CC=C1)C(O)=O)NC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)