Learn More
N,N-Dibutylformamide, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 112980100
| Quantity | 10mL |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C9H19NO | |
| MFCD00003289 | |
| dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide | |
| N,N-dibutylformamide |
Specifications
| N, N-Dibutylformamide | |
| 761-65-9 | |
| 100.0 | |
| 120.0°C (15.0 mmHg) | |
| 99+% | |
| C9H19NO | |
| HCON[(CH2)3CH3]2 | |
| MFCD00003289 | |
| 0.86 | |
| NZMAJUHVSZBJHL-UHFFFAOYSA-N | |
| N,N-dibutylformamide | |
| 12975 | |
| 99+% |
| 99+% | |
| 99.0 | |
| 0.8600g/mL | |
| 100°C | |
| Glass bottle | |
| 1.4430 to 1.4450 | |
| 10mL | |
| 03,443F | |
| dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide | |
| CCCCN(CCCC)C=O | |
| 157.26 | |
| 157.26 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses
GHS Signal Word: Danger
EINECSNumber : 212-090-