missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzyloxycarbonyl-L-cystine, 98%

Catalog No. AAH6391706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6391706 5 g
AAH6391714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6391706 Supplier Thermo Scientific Chemicals Supplier No. H6391706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6968-11-2 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6968-11-2
MDL Number MFCD00022030

Specifications

Melting Point 119°C
Color White
Molecular Formula C22H24N2O8S2
Beilstein 2550640
Synonym z-cys-oh 2, cbz-cys-oh 2, di-z-l-cystine, n,n'-dicarbobenzoxy-l-cystine, n,n'-di-cbz-l-cystine, 2r-2-benzyloxy carbonyl amino-3-2r-2-benzyloxy carbonyl amino-2-carboxyethyl disulfanyl propanoic acid, n,n'-dibenzyloxycarbonyl-l-cystine, n,n'-bis benzyloxycarbonyl-l-cystine, ambotzzaa1190, pubchem19022
InChI Key PTRQEEVKHMDMCF-UHFFFAOYNA-N
SMILES C1=CC=C(C=C1)COC(=O)NC(CSSCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)C(=O)O
IUPAC Name (2R)-3-[[(2R)-2-carboxy-2-(phenylmethoxycarbonylamino)ethyl]disulfanyl]-2-(phenylmethoxycarbonylamino)propanoic acid
Quantity 5 g
Molecular Weight (g/mol) 508.56
PubChem CID 385972
Formula Weight 508.56
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N,N'-Dibenzyloxycarbonyl-L-cystine
Show More Show Less
EINECSNumber 230-181-3
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only