missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dibenzylglycine Ethyl Ester 98.0+%, TCI America™
$188.27 - $749.35
Chemical Identifiers
| CAS | 77385-90-1 |
|---|---|
| Molecular Formula | C18H21NO2 |
| Molecular Weight (g/mol) | 283.371 |
| InChI Key | AFMDCFOWHWNQBP-UHFFFAOYSA-N |
| Synonym | Di-Bzl-Gly-OEt |
| PubChem CID | 3553452 |
| IUPAC Name | ethyl 2-(dibenzylamino)acetate |
| SMILES | CCOC(=O)CN(CC1=CC=CC=C1)CC2=CC=CC=C2 |
Chemical Identifiers
| 77385-90-1 | |
| 283.371 | |
| Di-Bzl-Gly-OEt | |
| ethyl 2-(dibenzylamino)acetate |
| C18H21NO2 | |
| AFMDCFOWHWNQBP-UHFFFAOYSA-N | |
| 3553452 | |
| CCOC(=O)CN(CC1=CC=CC=C1)CC2=CC=CC=C2 |
Specifications
| 77385-90-1 | |
| White | |
| 5 g | |
| AFMDCFOWHWNQBP-UHFFFAOYSA-N | |
| ethyl 2-(dibenzylamino)acetate | |
| 3553452 | |
| ≥98.0% (GC,N) | |
| N,N-Dibenzylglycine Ethyl Ester |
| 56°C | |
| C18H21NO2 | |
| Di-Bzl-Gly-OEt | |
| CCOC(=O)CN(CC1=CC=CC=C1)CC2=CC=CC=C2 | |
| 283.371 | |
| 283.37 | |
| Crystalline Powder |
Safety and Handling
TSCA : No