accessibility menu, dialog, popup

UserName

CAS RN 77385-90-1

H3COON

CAS RN 77385-90-1

IUPAC Name: ethyl 2-(dibenzylamino)acetate
Synonyms: ethyl 2-(dibenzylamino)acetate
Molecular Weight (g/mol): 283.37
Molecular Formula: C18H21NO2
InChi Key: AFMDCFOWHWNQBP-UHFFFAOYSA-N
SMILES: CCOC(=O)CN(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results