missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzylglycine Ethyl Ester 98.0+%, TCI America™

Catalog No. D31655G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31655G 5 g
D316525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D31655G Supplier TCI America Supplier No. D31655G
Only null left

Chemical Identifiers

CAS 77385-90-1
Molecular Formula C18H21NO2
Molecular Weight (g/mol) 283.371
InChI Key AFMDCFOWHWNQBP-UHFFFAOYSA-N
Synonym Di-Bzl-Gly-OEt
PubChem CID 3553452
IUPAC Name ethyl 2-(dibenzylamino)acetate
SMILES CCOC(=O)CN(CC1=CC=CC=C1)CC2=CC=CC=C2

Specifications

Melting Point 56°C
Color White
Quantity 5 g
Formula Weight 283.37
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N-Dibenzylglycine Ethyl Ester