missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine 98.0+%, TCI America™

Catalog No. D180725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D180725ML 25 mL
D1807500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D180725ML Supplier TCI America Supplier No. D180725ML

Chemical Identifiers

CAS 140-28-3
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00004771
InChI Key JUHORIMYRDESRB-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine, benzathine, dbed, benzatin, n1,n2-dibenzylethane-1,2-diamine, 1,2-bis benzylamino ethane, 1,2-ethanediamine, n,n'-bis phenylmethyl, usaf do-53, unii-c659vz7p7t, ethylenediamine, n,n'-dibenzyl
PubChem CID 8793
ChEBI CHEBI:51344
IUPAC Name N,N'-dibenzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Color White-Yellow
Boiling Point 175°C
Quantity 25 mL
UN Number 3259
Formula Weight 240.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material N,N′-Dibenzylethylenediamine
EINECSNumber (9)-0739
RTECSNumber KV3325000
TSCA Yes