missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine diacetate, 99%

CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol

Supplier:  Thermo Scientific Chemicals L1797436

 View more versions of this product

Catalog No. AAL1797436


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N,N'-Dibenzylethylenediamine diacetate
117°C to 118°C
MFCD00040588
3781362
n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2
360.454
360.46
122-75-8
C20H28N2O4
500 g
14,1064
MTRNNCLQPVCDLF-UHFFFAOYSA-N
acetic acid;N,N'-dibenzylethane-1,2-diamine
31228
99%

Chemical Identifiers

122-75-8
360.454
MTRNNCLQPVCDLF-UHFFFAOYSA-N
31228
CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2
C20H28N2O4
MFCD00040588
n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate
acetic acid;N,N'-dibenzylethane-1,2-diamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 204-572-4

RTECSNumber : KV3400000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only