Learn More
N,N'-Dibenzylethylenediamine diacetate, 99%
CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol
Supplier: Thermo Scientific Chemicals L1797422
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N,N'-Dibenzylethylenediamine diacetate | |
| 117°C to 118°C | |
| MFCD00040588 | |
| 3781362 | |
| n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate | |
| CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2 | |
| 360.454 | |
| 360.46 |
| 122-75-8 | |
| C20H28N2O4 | |
| 100 g | |
| 14,1064 | |
| MTRNNCLQPVCDLF-UHFFFAOYSA-N | |
| acetic acid;N,N'-dibenzylethane-1,2-diamine | |
| 31228 | |
| 99% |
Chemical Identifiers
| 122-75-8 | |
| 360.454 | |
| MTRNNCLQPVCDLF-UHFFFAOYSA-N | |
| 31228 | |
| CC(=O)O.CC(=O)O.C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2 |
| C20H28N2O4 | |
| MFCD00040588 | |
| n,n'-dibenzylethylenediamine diacetate, n1,n2-dibenzylethane-1,2-diamine diacetate, n,n'-dibenzyl ethylenediamine diacetate, benzathine diacetate, dbed diacetate, unii-x18b8x2c22, n,n'-dibenzylethylenediammonium di acetate, 1,2-ethanediamine, n,n'-bis phenylmethyl-, diacetate, bis acetic acid ; benzathine, 1,2-di benzylamino ethane diacetate | |
| acetic acid;N,N'-dibenzylethane-1,2-diamine |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 204-572-4
RTECSNumber : KV3400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only