missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine, 97%

Catalog No. AAL0354022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0354022 100 g
AAL0354014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0354022 Supplier Thermo Scientific Chemicals Supplier No. L0354022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 140-28-3 | C16H20N2 | 240.35 g/mol

N,N'-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Dibenzylethylenediamine is an intermediate of cephalosporin and penicillin.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Chemical Identifiers

CAS 140-28-3
Molecular Formula C16H20N2
Molecular Weight (g/mol) 240.35
MDL Number MFCD00004771
InChI Key JUHORIMYRDESRB-UHFFFAOYSA-N
Synonym n,n'-dibenzylethylenediamine, benzathine, dbed, benzatin, n1,n2-dibenzylethane-1,2-diamine, 1,2-bis benzylamino ethane, 1,2-ethanediamine, n,n'-bis phenylmethyl, usaf do-53, unii-c659vz7p7t, ethylenediamine, n,n'-dibenzyl
PubChem CID 8793
ChEBI CHEBI:51344
IUPAC Name N,N'-dibenzylethane-1,2-diamine
SMILES C1=CC=C(C=C1)CNCCNCC2=CC=CC=C2

Specifications

Melting Point 23°C to 25°C
Density 1.025
Boiling Point 184°C to 186°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.565
Linear Formula C6H5CH2NHCH2CH2NHCH2C6H5
Quantity 100 g
UN Number UN2735
Beilstein 786668
Sensitivity Air Sensitive
Merck Index 14,1064
Solubility Information Slightly soluble in water.
Formula Weight 240.35
Percent Purity 97%
Chemical Name or Material N,N'-Dibenzylethylenediamine
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 205-408-4
RTECSNumber KV3325000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only