missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-tert-butylcarbodiimide 98.0+%, TCI America™

Catalog No. p-7128744
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53475ML 5 mL
D534725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53475ML Supplier TCI America Supplier No. D53475ML
Only null left

Chemical Identifiers

CAS 691-24-7
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.257
MDL Number MFCD00010231
InChI Key IDVWLLCLTVBSCS-UHFFFAOYSA-N
PubChem CID 69650
IUPAC Name N,N'-ditert-butylmethanediimine
SMILES CC(C)(C)N=C=NC(C)(C)C

Specifications

Color Colorless
Boiling Point 50°C
Quantity 5 mL
UN Number 1993
Formula Weight 154.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N′-Di-tert-butylcarbodiimide