missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-octylaniline, 98%

Catalog No. AAH2664406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2664406 5 g
AAH2664403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2664406 Supplier Thermo Scientific Chemicals Supplier No. H2664406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3007-75-8 | C22H39N | 317.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3007-75-8
Molecular Formula C22H39N
Molecular Weight (g/mol) 317.56
MDL Number MFCD00155305
InChI Key PTRSTXBRQVXIEW-UHFFFAOYSA-N
Synonym n,n-di-n-octylaniline, benzenamine, n,n-dioctyl, dioctylphenylamine, benzenamine,n,n-dioctyl
PubChem CID 137791
IUPAC Name N,N-dioctylaniline
SMILES CCCCCCCCN(CCCCCCCC)C1=CC=CC=C1

Specifications

Refractive Index 1.5495
Quantity 5 g
Formula Weight 317.56
Percent Purity 98%
Chemical Name or Material N,N-Di-n-octylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only