accessibility menu, dialog, popup

UserName

CAS RN 3007-75-8

IUPAC Name: N,N-dioctylaniline
Synonyms: N,N-dioctylaniline
Molecular Weight (g/mol): 317.56
Molecular Formula: C22H39N
InChi Key: PTRSTXBRQVXIEW-UHFFFAOYSA-N
SMILES: CCCCCCCCN(CCCCCCCC)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results