missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-butylthiourea, 98%

Catalog No. p-7023343
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1678318 50 g
AAA1678330 250 g
AAA167830B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1678318 Supplier Thermo Scientific Chemicals Supplier No. A1678318
Only null left

CAS: 109-46-6 | C9H20N2S | 188.333 g/mol

N,N'-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylthiourea is used in vulcanization accelerator and corrosion inhibitors for metal treatments. It is used as leather, plastic and rubber auxiliary agents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 109-46-6
Molecular Formula C9H20N2S
Molecular Weight (g/mol) 188.333
MDL Number MFCD00004926
InChI Key KFFQABQEJATQAT-UHFFFAOYSA-N
Synonym 1,3-dibutyl-2-thiourea, n,n'-dibutylthiourea, thiourea, n,n'-dibutyl, pennzone b, n,n'-di-n-butylthiourea, thiate u, 1,3-di-n-butyl-2-thiourea, urea, 1,3-dibutyl-2-thio, usaf ek-2138, urea, 1,3-di-n-butyl-2-thio
PubChem CID 2723622
IUPAC Name 1,3-dibutylthiourea
SMILES CCCCNC(=S)NCCCC

Specifications

Melting Point 64°C to 67°C
Odor Odorless
Quantity 50 g
Beilstein 507434
Solubility Information Insoluble in water.
Formula Weight 188.34
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylthiourea
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 203-674-6
RTECSNumber YS8400000
TSCA Yes

RUO – Research Use Only