missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-butylformamide, 99%

CAS: 761-65-9 | C9H19NO | 157.26 g/mol

Supplier:  Thermo Scientific Chemicals L1275522

 View more versions of this product

Catalog No. AAL1275522


Only null left
Explore available promotions

Description

Description

N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

N,N-Di-n-butylformamide
<-30°C
119°C to 120°C (15 mmHg)
C9H19NO
MFCD00003289
636100
Insoluble in water.
CCCCN(CCCC)C=O
157.26
157.26
761-65-9
0.864
115°C (239°F)
1.4445
100 g
dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
NZMAJUHVSZBJHL-UHFFFAOYSA-N
N,N-dibutylformamide
12975
99%

Chemical Identifiers

761-65-9
157.26
NZMAJUHVSZBJHL-UHFFFAOYSA-N
12975
CCCCN(CCCC)C=O
C9H19NO
MFCD00003289
dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
N,N-dibutylformamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H302-H311-H315-H319

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 212-090-0

RTECSNumber : LQ1833000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only