missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-butylformamide, 99%

Catalog No. AAL1275514
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1275514 25 g
AAL1275522 100 g
AAL1275536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1275514 Supplier Thermo Scientific Chemicals Supplier No. L1275514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 761-65-9 | C9H19NO | 157.26 g/mol

N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Di-n-butylformamide is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 761-65-9
Molecular Formula C9H19NO
Molecular Weight (g/mol) 157.26
MDL Number MFCD00003289
InChI Key NZMAJUHVSZBJHL-UHFFFAOYSA-N
Synonym dibutylformamide, formamide, n,n-dibutyl, n,n-di-n-butylformamide, formamide, n,n-di-n-butyl, dibutylamid kyseliny mravenci, dibutylamid kyseliny mravenci czech, formamide, dibutyl, n-formyldibutylamine, formamide,n-dibutyl, n,n-di-butylformamide
PubChem CID 12975
IUPAC Name N,N-dibutylformamide
SMILES CCCCN(CCCC)C=O

Specifications

Melting Point <-30°C
Density 0.864
Boiling Point 119°C to 120°C (15 mmHg)
Flash Point 115°C (239°F)
Refractive Index 1.4445
Quantity 25 g
Beilstein 636100
Solubility Information Insoluble in water.
Formula Weight 157.26
Percent Purity 99%
Chemical Name or Material N,N-Di-n-butylformamide
Show More Show Less
Hazard Category H302-H311-H315-H319
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 212-090-0
RTECSNumber LQ1833000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only