missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D49291G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49291G 1 g
D49295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49291G Supplier TCI America Supplier No. D49291G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 139994-47-1
Molecular Formula C38H28N2
Molecular Weight (g/mol) 512.656
MDL Number MFCD28291914
InChI Key QVDYERLGSGAPKP-UHFFFAOYSA-N
PubChem CID 18938637
IUPAC Name 1-N,4-N-dinaphthalen-2-yl-1-N,4-N-diphenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=CC=C3)C4=CC5=CC=CC=C5C=C4)C6=CC7=CC=CC=C7C=C6

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 1 g
Formula Weight 512.66
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N,N′-Di(2-naphthyl)-N,N′-diphenyl-1,4-phenylenediamine