CAS RN 139994-47-1
IUPAC Name:
N1,N4-bis(naphthalen-2-yl)-N1,N4-diphenylbenzene-1,4-diamine
Synonyms:
N1,N4-bis(naphthalen-2-yl)-N1,N4-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol):
512.66
Molecular Formula:
C38H28N2
InChi Key:
QVDYERLGSGAPKP-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)N(C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1)C1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
N,N'-Di(2-naphthyl)-N,N'-diphenyl-1,4-phenylenediamine 98.0+%, TCI America™
CAS: 139994-47-1 Molecular Formula: C38H28N2 Molecular Weight (g/mol): 512.656 MDL Number: MFCD28291914 InChI Key: QVDYERLGSGAPKP-UHFFFAOYSA-N PubChem CID: 18938637 IUPAC Name: 1-N,4-N-dinaphthalen-2-yl-1-N,4-N-diphenylbenzene-1,4-diamine SMILES: C1=CC=C(C=C1)N(C2=CC=C(C=C2)N(C3=CC=CC=C3)C4=CC5=CC=CC=C5C=C4)C6=CC7=CC=CC=C7C=C6