missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)ethylenediamine, 97%

Catalog No. AAL1343706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1343706 5 g
AAL1343714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1343706 Supplier Thermo Scientific Chemicals Supplier No. L1343706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 19522-69-1 | C6H16N2 | 116.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19522-69-1
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00025602
InChI Key DFPGBRPWDZFIPP-UHFFFAOYSA-N
Synonym n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
PubChem CID 519668
IUPAC Name N'-butylethane-1,2-diamine
SMILES CCCCNCCN

Specifications

Density 0.836
Boiling Point 170°C to 171°C
Flash Point 59°C (138°F)
Refractive Index 1.443
Quantity 5 g
UN Number UN2734
Beilstein 1732700
Sensitivity Air Sensitive
Formula Weight 116.21
Percent Purity 97%
Chemical Name or Material N-(n-Butyl)ethylenediamine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only