accessibility menu, dialog, popup

UserName

CAS RN 19522-69-1

IUPAC Name: (2-aminoethyl)(butyl)amine
Synonyms: (2-aminoethyl)(butyl)amine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: DFPGBRPWDZFIPP-UHFFFAOYSA-N
SMILES: CCCCNCCN
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)