missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%

CAS: 14167-18-1 | C16H14CoN2O2 | 325.23 g/mol

Supplier:  Thermo Scientific Chemicals 04317406

 View more versions of this product

Catalog No. AA4317406


Only null left
Explore available promotions

Description

Description

Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N,N'-Bis (salicylidene)ethylenediaminecobalt (II)
Black
C16H14CoN2O2
MFCD00000009
Salcomine
NPAQSKHBTMUERN-UHFFFAOYSA-L
cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
57448789
96%
14167-18-1
Odorless
[-CH2N=CH(C6H4O-)]2Co
5 g
Insoluble in water. Soluble in benzene,chloroform,and pyridine.
[Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
325.23
325.23
Powder

Chemical Identifiers

14167-18-1
325.23
NPAQSKHBTMUERN-UHFFFAOYSA-L
57448789
C16H14CoN2O2
MFCD00000009
Salcomine
[Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c

H302+H332-H317-H350

EINECSNumber : 238-012-5

RTECSNumber : GG0590000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only