missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediaminecobalt(II), 96%

Catalog No. p-7058735
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4317403 1 g
AA4317406 5 g
AA4317414 25 g
AA4317422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4317403 Supplier Thermo Scientific Chemicals Supplier No. 04317403
Only null left

CAS: 14167-18-1 | C16H14CoN2O2 | 325.23 g/mol

Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Molecular Weight (g/mol) 325.23
MDL Number MFCD00000009
InChI Key NPAQSKHBTMUERN-UHFFFAOYSA-L
Synonym Salcomine
PubChem CID 57448789
SMILES [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]

Specifications

Color Black
Odor Odorless
Linear Formula [-CH2N=CH(C6H4O-)]2Co
Quantity 1 g
Solubility Information Insoluble in water. Soluble in benzene,chloroform,and pyridine.
IUPAC Name cobalt;(6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
Formula Weight 325.23
Percent Purity 96%
Physical Form Powder
Chemical Name or Material N,N'-Bis (salicylidene)ethylenediaminecobalt (II)
Hazard Category H302+H332-H317-H350
Hazard Statement GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
EINECSNumber 238-012-5
RTECSNumber GG0590000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only